Pierce™ Streptavidin UltraLink™ Resin, Thermo Scientific
Supplier: Thermo Scientific
53113 53114
CAPI53113EA 850.58 CAD
CAPI53113 CAPI53114
Pierce™ Streptavidin UltraLink™ Resin, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ Streptavidin UltraLink™ resin consists of purified recombinant streptavidin that has been covalently immobilized onto a beaded acrylamide support
  • Acrylamide based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
  • Streptavidin UltraLink™ resin binding capacity: >2 mg of biotinylated BSA/mL of resin
UltraLink™ beads are hydrophilic, charge free, highly crosslinked, rigid and durable which makes the affinity resin suitable for a variety of batch or column type affinity procedures that require medium pressure chromatography.
Caution: For Research Use Only. Not for use in diagnostic procedures.