Pierce™ DyLight™ Antibody Labelling Dyes, Thermo Scientific
Supplier: Thermo Scientific
46400 46401 46402 46403 46412 46413 46414 46417 46418 46419 46421 46422 46600 46602 46608 46613 46618 46621 62262 62263 62265 62266 62290 62295
CAPI46400EA 1037.54 CAD
CAPI46400 CAPI46401 CAPI46402 CAPI46403 CAPI46412 CAPI46413 CAPI46414 CAPI46417 CAPI46418 CAPI46419 CAPI46421 CAPI46422 CAPI46600 CAPI46602 CAPI46608 CAPI46613 CAPI46618 CAPI46621 CAPI62262 CAPI62263 CAPI62265 CAPI62266 CAPI62290 CAPI62295
Pierce™ DyLight™ Antibody Labelling Dyes, Thermo Scientific
Protein Linking and Labeling Systems
DyLight™ Dyes have absorption spectra ranging from 350 nm to 770 nm and match the output wavelengths of common fluorescence instrumentation
  • Available in both amine- and sulfhydryl-reactive chemistries for fast and efficient labeling of IgG or other proteins
  • High water solubility
  • Excellent photostability
  • Compatible with common fluorescence instrumentation
Additionally, the water solubility of the DyLight™ reagents allows a high dye-to-protein ratio without precipitation during conjugation.
The DyLight™ dyes exhibit higher fluorescence intensity and photostability than Alexa Fluor®, CyDye® and LI-COR dyes in many applications and remain highly fluorescent over a broad pH range (pH 4 to 9). Additionally, the water solubility of the DyLight™ reagents allows a high dye-to-protein ratio without precipitation during conjugation.
Delivery information: Supplied as 1 mg or in convenience packs containing sufficient reagent for 5 single labelling reactions. Each single-use vial is sufficient to label 0.25 to 1 mg of protein (>45 kDa).
Caution: For Research Use Only. Not for use in diagnostic procedures.