Pierce™ Protein A Spin Plate for IgG Screening, Thermo Scientific
Supplier: Thermo Scientific
45202 45204
CAPI45202EA 722.83 CAD
CAPI45202 CAPI45204
Pierce™ Protein A Spin Plate for IgG Screening, Thermo Scientific
Microplates Immunology Microplates and Strips
Pierce™ Protein A and Protein G Spin Plates for IgG Screening offers a high throughput format for quick purification and screening of antibodies and for immunoprecipitation (IP)
  • Immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
  • Agarose resin
  • Inert and stable
  • High capacity
The Protein A and Protein G resin purifies by binding IgGs, primarily through their Fc region, in a buffer that facilitates the interaction. Non-relevant proteins, such as albumin and transferrin, are removed by washing and the purified antibody is then recovered using low-pH elution conditions.
Each well of the 96-well spin plate can process 10 to 100 µl of serum, cell culture supernatant or ascites fluid samples.
Delivery information: Each pack contains 1 plate (each well contains 50 µl of resin for processing 10 to 100 µl of sample) and 3 wash/collection plates.
Caution: For Research Use Only. Not for use in diagnostic procedures.