Pierce™ Mouse IgG1 Fab and F(ab')2 Preparation Kit, Thermo Scientific
Supplier: Thermo Scientific
The Thermo Scientific Pierce Mouse IgG1 Fab and F(ab')2 Micro Preparation Kit uses immobilized ficin protease to efficiently digest mouse IgG1 into Fab or F(ab')2 fragments, depending on the chosen reaction conditions.
- Capacity—kit provides for fragmenting 25 to 250 µg IgG.
- Complete—kit includes all reagents needed to prepare and purify antibody fragments
- Enzyme-free digestion products—Immobilized Ficin (beaded agarose resin) provides for control of the digestion reaction and complete removal of resulting antibody fragments from the proteolytic enzyme
The kit procedure is optimized for mouse IgG1, and is the only effective method for this IgG species and subclass.
Ordering information: Sufficient For: 10 antibody samples, each containing 25 to 250 µg IgG
Pierce Ficin Agarose, 0.8 mL
Cysteine-HCl, 1 g
Mouse IgG1 Digestion Buffer, 120 mL
NAb Protein A Plus Spin Columns, 0.2 mL, 2 columns
Protein A Binding Buffer, 50 mL
IgG Elution Buffer, 50 mL
Zeba Spin Desalting Columns, 7K MWCO, 0.5 mL, 10 columns
Spin Columns, 0.8 mL, 10 columns
Microcentrifuge tubes, 2 mL, 30 tubes
Store at 4 – 8 °C.
Delivery information: Kit includes ficin agarose (2.5 mL), cysteine-HCl (1 g), mouse IgG1 digestion buffer (120 mL), NAb™ Protein A Plus spin column (1 x 1 mL), Protein A binding buffer (120 mL), IgG elution buffer (120 mL), Zeba desalt spin columns, 7K MWCO (10 x 2 mL), spin columns (10 x 0.8 mL) and microcentrifuge tubes(30 x 2 mL).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...