Alkaline Phosphatase

Supplier: Thermo Scientific

31391 31392
CAPI31391EA 2859.77 CAD
CAPI31391 CAPI31392
Alkaline Phosphatase
Enzymes

This purified calf intestinal alkaline phosphatase (CIP) is supplied in Tris buffer and 50% glycerol for use in protein research methods.


  • Purified form, ready to dilute and conjugate without prior dialysis
  • Concentrated approximately 20 mg/mL
  • High specific activitywhich is typically greater than 1600 units/mg
  • Tris buffered solution supplied in 5 mM Tris, 5 mM magnesium chloride and 0.1 mM zinc chloride
  • pH ~7.0 in 50% glycerol


The main applications for alkaline phosphatase in molecular biology and protein research are to remove 5'-phosphate groups from DNA or as a reporter system for immunoassays such as ELISA


One unit equals the amount of protein required to hydrolyze one micromole of p-nitrophenyl phosphate (PNPP) per minute at 25°C in a glycine buffer, pH 9.6.


For these latter methods, the enzyme is usually conjugated to specific primary or secondary antibodies and its activity is detected with a color-forming (or light-generating) substrate.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now

SPECIFICATIONS

Enzyme name Alkaline Phosphatase
Environmentally Preferable
Source
Species
Storage conditions 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR