Pierce™ Wash Buffers, Thermo Scientific
Supplier: Thermo Scientific
28341
28344
28346
28348
28352
28358
28360
CAPI28341EA
247.28
CAD
CAPI28341
CAPI28344
CAPI28346
CAPI28348
CAPI28352
CAPI28358
CAPI28360
Pierce™ Wash Buffers, Thermo Scientific
Buffers
Electrophoresis Buffers
Washing Buffers
These wash buffers are designed for use in dialysis, cross-linking, enzyme assays, ELISAs, immunohistochemistry, protein plate-coating, biotinylation applications.
- Ready to use without having to reconstitute with ultrapure water
- Eliminates the possibility of powder remaining in a packet
- 10X or 20X concentration eliminates time spent waiting for powder to dissolve
- Storage as concentrated stock minimizes bench space needed for large volume solutions
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25˚C.
Caution: For Research Use Only. Not for use in diagnostic procedures.