N-Octyl-β-D-glucopyranoside non-ionic detergent, Pierce™

Supplier: Thermo Scientific
Warning

Synonyms: Octylglucopyranoside1-O-n-Octyl-β-D-glucopyranosideOctyl-β-D-glucopyranosiden-Octyl β-D-glucopyranosideOG(2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol

28310 28310
CAPI28310EA 1382.94 CAD
CAPI28310 PI28310
N-Octyl-β-D-glucopyranoside non-ionic detergent, Pierce™
N-Octyl-β-D-glucopyranoside

Thermo Scientific Octyl-beta-Glucoside is a nonionic detergent that has been widely used for membrane protein solubilization.


  • A low-molecular weight, nonionic detergent
  • Effective for membrane protein solubilization
  • Less stable and less commonly used than octylthioglucoside (OTG)
  • Can be removed from solution by dialysis
  • Optically clear; low absorbance at 280 nm


White, free-flowing powder, free of foreign material. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: C₁₄H₂₈O₆
MW: 292.37 g/mol
Melting Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00063288
CAS Number: 29836-26-8
EINECS: 249-887-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR