Pierce™ PEGylated SPDP Crosslinkers, Thermo Scientific
Supplier: Thermo Scientific
26128 26129
CAPI26128EA 1257.65 CAD
CAPI26128 CAPI26129
Pierce™ PEGylated SPDP Crosslinkers, Thermo Scientific
Protein Linking and Labeling Systems
Thermo Scientific Pierce™ PEGn-SPDP is a multifunctional crosslinker for protein conjugation via amine-to-amine or amine-to-sulfhydryl crosslinks that contain a n-unit polyethylene glycol (PEG) group and a reducible (cleavable) disulfide bond.
  • Efficient amine conjugation—NHS ester provides effective conjugation to lysines, N-termini of peptides and other primary amines
  • Reversible sulfhydryl conjugation—pyridyldithiol group enables disulfide linkage with reduced cysteines and other sulfhydryl groups, providing a stable but cleavable bond
  • Polyethylene glycol—PEG groups are flexible, non-immunogenic, hydrophilic, and often enhance the solubility of attached molecules
  • Spacer arm—connects primary amines and sulfhydryl targets
  • Upon receipt store at -20˚C protected from moisture
SPDP-type reagents have an amine-reactive N-hydroxysuccinimide (NHS) ester at one end and a suflhydryl-reactive 2-pyridyldithiol group at the opposite end. PEGn-SPDP has a n-unit polyethylene glycol spacer arm, which confers greater solubility to the crosslinker and linked proteins compared to crosslinkers having only hydrocarbon spacers. Pyridyldithiol reagents produce disulfide-containing linkages that can be cleaved with reducing agents such as dithiothreitol (DTT). Crosslinking experiments with SPDP reagents are not limited to those involving proteins. Any of a variety of molecules with primary amines and sulfhydryl groups can be modified or crosslinked using an SPDP reagent.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable
Host
Learn more