Pierce™ CA(PEG)n Carboxy-PEG-Amine Compound, Thermo Scientific
Supplier: Thermo Scientific
The carboxy-PEGn-amine (CA[PEG]n) PEGylation reagents are zwitterionic, amino acid derivatives that are used for modifying proteins or surfaces such as beads, nanoparticles and self-assembled monolayers
- Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
- Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
- PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
- Allows site-specific labeling or crosslinking involving primary amines or carboxyl groups on proteins or surfaces
- Easy-to-follow instructions increase the likelihood of a successful outcome
CA(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with carboxy (-COOH) and amino (-NH2) termini. The unbranched, hydrophilic, discrete-length molecules have the form Carboxy-PEGn-Amine, where the subscript "n" denotes 4, 8, 12, or 24 ethylene glycol units. The carboxyl and primary amine of each compound provide specific targets for crosslinking and other conjugation methods, making these compounds useful as PEGylation reagents.
These PEGylation reagents are homogenous compounds of defined molecular weight and spacer length, providing precision in optimizing modification applications.
Ordering information: Store desiccated at 4?C. Shipped at ambient temperature.
Caution: For Research Use Only. Not for use in diagnostic procedures.