Pierce™ Glutathione Superflow Agarose Affinity Chromatography Media, Thermo Scientific
Supplier: Thermo Scientific
25236 25237 25238
CAPI25236EA 791.29 CAD
CAPI25236 CAPI25237 CAPI25238
Pierce™ Glutathione Superflow Agarose Affinity Chromatography Media, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Pierce Glutathione Superflow Agarose consists of glutathione that has been immobilized to a crosslinked 6% beaded agarose that is designed for high-purity, large-scale purification of GST-tagged proteins.
  • High dynamic binding capacity—binds 10 mg of pure GST per mL of resin at a linear flow rate of 150 cm/hour; can bind greater than 20 mg/mL at 30 cm/hour flow rate
  • High purity—provides greater than 90% purity when used to purify overexpressed GST or GST-fusion proteins from lysates
  • Robust—highly crosslinked 6% agarose Superflow beaded support tolerates linear flow rates up to 1200 cm/hour
  • Compatible—maintains function after exposure to a wide variety of chemicals and pH values
  • Cost effective—resin is stable through multiple cycles of cleaning and reuse
Glutathione Superflow Agarose is a highly crosslinked, durable resin that does not compress at flow rates required for medium- to large-scale FPLC purifications. The resin holds up well to a variety of chemical and pH values, and it is compatible with common clean-in-place procedures. The specific binding of glutathione to the GST-tag results in high purity and high yield in a single step purification process.
Caution: For Research Use Only. Not for use in diagnostic procedures.