Pierce™ Protein A Plus Agarose, Thermo Scientific
Supplier: Thermo Scientific
22810 22811 22812
CAPI22810EA 357.15 CAD
CAPI22810 CAPI22811 CAPI22812
Pierce™ Protein A Plus Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Pierce Protein A Plus agarose is a high capacity Protein A beaded agarose resin for use in a variety of antibody affinity purification methods.
  • Immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein A is immobilized by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • High capacity beads with a dense load of immobilized Protein A
Caution: For Research Use Only. Not for use in diagnostic procedures.