Pierce™ Pegylation Reagents, MS(PEG)n Methyl-PEG-NHS-Esters, Thermo Scientific
Supplier: Thermo Scientific
22341 22342 22424 22509 22685 22686 22687
CAPI22341EA 701.19 CAD
CAPI22341 CAPI22342 CAPI22424 CAPI22509 CAPI22685 CAPI22686 CAPI22687
Pierce™ Pegylation Reagents, MS(PEG)n Methyl-PEG-NHS-Esters, Thermo Scientific
Protein Purification Protein Purification Reagents
Thermo Scientific Pierce MS(PEG)n reagents are methyl-terminated, polyethylene glycol compounds (n equals 4 to 24 PEG units) and TMS(PEG)12 is a three-branched, polyethylene glycol compound (3 x 12 PEG units). Both are activated as NHS esters for covalent pegylation of primary amines on proteins (eg. lysines) or assay surfaces.
  • NHS-activated for efficient PEGylation of primary amines at pH 7-9; reaction of NHS-ester group results in formation of stable, irreversible amide bonds
  • Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
  • Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
  • PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Easy-to-follow instructions increase the likelihood of a successful outcome
MS(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with methyl (—CH3) and amine-reactive NHS-ester groups at opposite ends
TMS(PEG)12 is the abbreviation for a branched trimethyl (TM) and succinimide ester (S) derivative of polyethylene glycol (PEG) for efficient and specific modification of primary amines. Each methyl-terminated PEG (mPEG) branch contains 12 ethylene glycol units. The three branches are attached to a 4-unit PEG stem that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at the distal end. The NHS ester is spontaneously reactive with primary amines (—NH2), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.
The unbranched, hydrophilic, discrete-length molecules have the form Methyl-PEGn-NHS Ester, where the subscript "n" denotes 4, 8, 12, or 24 ethylene glycol units. The N-hydroxysuccinimide (NHS) ester is spontaneously reactive with primary amines (—NH2), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage conditions Store desiccated at -20˚C.