Amine Reactive Biotinylation Kits, EZ-Link™, Thermo Scientific
Supplier: Thermo Scientific
21425 21435 21445 21455 21925 21935 21945 21955
CAPI21425EA 1049.14 CAD
CAPI21425 CAPI21435 CAPI21445 CAPI21455 CAPI21925 CAPI21935 CAPI21945 CAPI21955
Amine Reactive Biotinylation Kits, EZ-Link™, Thermo Scientific
Protein Linking and Labeling Systems
The amine group is the most common target for modifying protein molecules because it is abundant in the majority of proteins either due to the presence of lysine bearing amino side chain functionality or the N-terminal-amine. Amine-reactive biotinylation reagents can be divided into two groups based on water solubility: NHS-esters and sulfo-NHS-esters.
NHS-esters of biotin are water-insoluble and membrane-permeable because they do not possess a charged group
Sulfo-NHS-esters are water soluble and useful for applications that cannot tolerate organic solvents. Sulfo-NHS-esters of biotin are recommended for use as cell surface biotinylation reagents. Because of the charged sulfonate group, these compounds do not penetrate the plasma membrane, thus sulfo-NHS-esters biotinylation is restricted to the cell surface.
EZ-Link™ Sulfo-NHS Biotinylation Kits: Complete kit with an optimized protocol for labeling a protein and determining how much biotin has been attached. Fast and efficient labeling procedure.
EZ-Link™ Sulfo-NHS-LC Biotinylation Kits: Complete kit with an optimized protocol for labeling a protein and determining how much biotin has been attached. Fast and efficient labeling procedure.
EZ-Link™ NHS-PEG4 Biotinylation Kits: PEG4-Biotin transfers water-solubility to biotinylated molecules.
EZ-Link™ Sulfo-NHS-SS Biotinylation Kits: Cleavable biotinylation reagent allows for later removal of the biotin label.
They may be used for biotinylating internal as well as external cellular components. Since NHS-esters of biotin are water-insoluble, for reactions in aqueous solution they must first be dissolved in an organic solvent, such as DMSO and DMF.
Caution: For Research Use Only. Not for use in diagnostic procedures.