Pierce™ High Sensitivity Streptavidin-HRP, Thermo Scientific
Supplier: Thermo Scientific
21130 21134
CAPI21130EA 553.34 CAD
CAPI21130 CAPI21134
Pierce™ High Sensitivity Streptavidin-HRP, Thermo Scientific
Protein Purification Protein Purification Reagents
Thermo Scientific Pierce High Sensitivity Streptavidin-HRP is an exclusive peroxidase-conjugated streptavidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.
  • Detect low levels of target without background; obtain high signal-to-noise ratios
  • Less conjugate in Western blotting and ELISA applications and still obtain excellent results
  • Compatible with typical chemiluminescent, fluorescent and colorimetric peroxidase substrates
  • Ready-to-use stabilized liquid format means there is no waiting for thawing and no need to aliquot
  • It can be diluted 8-to 10-fold more than traditional HRP conjugates and still produce similar results
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage conditions Storage at 4˚C for at least one year.