Restore™ Western Blot Stripping Buffer, Thermo Scientific
Supplier: Thermo Scientific
21059 21062 21063
CAPI21059EA 415.13 CAD
CAPI21059 CAPI21062 CAPI21063
Restore™ Western Blot Stripping Buffer, Thermo Scientific
Buffers Electrophoresis Buffers Stripping Buffers
Thermo Scientific Restore™ Western Blot Stripping Buffer safely and effectively removes primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.
  • Saves time—no need to re-run gels and blots
  • Saves costly sample—re-probe the membrane using the same target sample
  • Effective—formulation is more efficient at stripping antibodies than homemade buffers
  • Gentle—does not damage the target antigen during stripping allowing efficient reprobing
  • Odor-free—no mercaptans means no acrid odors
  • Economical—less expensive than other commercial stripping buffers
Performing gel electrophoresis and duplicate immunoblot assays to test new primary antibodies or antibody concentrations is time-consuming and expensive. Restore™ Western blot stripping buffer offers an effective alternative.
Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at room temperature or 4˚C. Product shipped at ambient temperature.
Order Now
SPECIFICATIONS
Environmentally Preferable