Pierce™ Protein L Agarose, Thermo Scientific
Supplier: Thermo Scientific
20510 20512
CAPI20510EA 896.42 CAD
CAPI20510 CAPI20512
Pierce™ Protein L Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Pierce Protein L Agarose is a premium-quality affinity resin for antibody purification.
  • Immobilized Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein L is immobilized by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • Standard capacity beads with a normal load of immobilized Protein L
Pierce Protein L Agarose consists of recombinant Protein L that has been covalently immobilized onto high-quality crosslinked 6% beaded agarose (CL-6B)
Pierce Protein L Agarose is a versatile affinity resin for purification of mammalian IgG isotypes that contain specific kappa light chains. Protein L is especially suited for isolating mouse IgG VkI and human IgG VkI, VkIII and VkIV monoclonal antibodies.
This particular variety of the resin provides the most versatile combination of chromatographic features for high yield and high purity purification of target immunoglobulins. The agarose beads have physical and chemical properties suitable for many affinity purification systems.
Delivery information: Product is shipped at ambient temperature.
Caution: For Research Use Only. Not for use in diagnostic procedures. Store product at 4-80˚C
Order Now
SPECIFICATIONS
Environmentally Preferable
Binding capacity 5–10 mg Human IgG/mL Resin