Pierce™ Detoxi-Gel™ Endotoxin Removing Gel and Columns, Thermo Scientific
Supplier: Thermo Scientific
20339 20344
CAPI20339EA 584.96 CAD
CAPI20339 CAPI20344
Pierce™ Detoxi-Gel™ Endotoxin Removing Gel and Columns, Thermo Scientific
Protein Extraction Protein Extraction Kits
Thermo Scientific Detoxi-Gel Endotoxin Removing Gel uses Polymixin B ligand immobilized on beaded affinity resin to bind and extract endotoxins from antibody or protein samples.
  • Resin uses immobilized polymixin B to remove pyrogens by binding their lipid A domains
  • Reduces endotoxin levels in protein solutions, cell culture media, solutions containing pharmacological components and aqueous buffers
  • 1 mL of gel removes>9995 EU from a 10,000 EU challenge of LPS (greater than 99% efficiency)
  • 1 mL of gel binds 4000-6000 E.U. of the lipopolysaccharide from E. coli strain 055:BS
  • Protein recovery dependent upon protein type and concentration; some empirical testing required
  • Stable and reusable; resin can be regenerated by stripping off the endotoxins with a 1% deoxycholate solution in pyrogen-poor water; no loss in binding capacity even after 10 regenerations
Detoxi-Gel Endotoxin Removing Gel and quickly remove endotoxins (lipopolysaccharides or LPS) from samples. Even low levels of endotoxins can be toxic to cells or organisms and must be removed before biological samples can be introduced.
Caution: Store at 4˚C. For Research Use Only. Not for use in diagnostic procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable