Pierce™ Monomeric Avidin Agarose, Thermo Scientific
Supplier: Thermo Scientific
20228 20267
CAPI20228EA 1442.44 CAD
CAPI20228 CAPI20267
Pierce™ Monomeric Avidin Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Pierce Monomeric Avidin Agarose is ideal for purifying biotinylated proteins, peptides and other molecules.
  • Non-denaturing—purifies biotinylated products using mild elution conditions (2 mM free biotin)
  • Reusable—Monomeric Avidin Agarose can be regenerated and reused at least 10 times, with only a marginal loss in biotin binding capacity (approximately 2.5% decrease per regeneration)
  • Specific—retains biotin-binding specificity and low nonspecific binding of native avidin
  • Flexible—bind samples in a variety of physiological buffers and elute either with 0.1M glycine or by competition with 2 mM biotin
Caution: For Research Use Only. Not for use in diagnostic procedures.