Pierce™ Anti-c-Myc Agarose, Thermo Scientific
Supplier: Thermo Scientific
20168 20169
CAPI20168EA 1333.95 CAD
CAPI20168 CAPI20169
Pierce™ Anti-c-Myc Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Pierce Anti-c-Myc Agarose is a high-affinity immunoprecipitation resin ideal for purification of recombinant c-Myc-tagged proteins expressed in human in vitro expression systems and bacterial and mammalian cell lysates.
  • Highly specific anti-c-Myc monoclonal antibody (clone 9E10) enables high yield and high purity for immunoprecipitation
  • Available in 2 and 10 mL resin package sizes to allow for larger scale purifications or immunoprecipitations
  • Can be used in spin or gravity columns as well as in FPLC cartridges
  • Reagents to elute and detect c-Myc tagged fusion proteins are available separately
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage conditions Store at 4˚C. Do not freeze.