Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer, Thermo Scientific
Supplier: Thermo Scientific
Thermo Scientific Pierce Phenol-Chloroform is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1).
- Offers excellent, nuclease-free performance
- Supplied at pH 6.7 for special, lower pH DNA extractions; for most DNA applications, the alkaline buffer (included) should be added to produce a pH of 8.0
- Stored at 2 to 8 °C
This buffer is ideal for use in applications of DNA extraction and purification.
Ordering information:
Caution: For Research Use Only. Not for use in diagnostic procedures.