Formamide ≥99.5%, high purity

Supplier: Thermo Scientific

Synonyms: Carbamaldehyde

17899 17899
CAPI17899EA 287.53 CAD
PI17899 CAPI17899
Formamide ≥99.5%, high purity
Formamide

Thermo Scientific Pierce Formamide is a high-purity formulation of formamide denaturant that is suitable for the preparation of hybridization cocktails and various denaturing solutions used with nucleic acids.


  • Ultra pure formulation possesses a high degree of purity and is free of contaminating RNase and DNase
  • Will not cause degradation of expensive or hard to isolate RNA and DNA samples
  • Pre-filtered through a mixed-ion exchange resin to eliminate products of decomposition and residual metal cations
  • Storage Conditions: 2 to –8 °C


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: HCONH₂
MW: 45.04 g/mol
MDL Number: MFCD00007941
CAS Number: 75-12-7
https://ca.vwr.com/stibo/web/std.lang.all/25/02/2302502.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR