Pierce™ Firefly Luciferase Flash Assay Kit, Thermo Scientific
Supplier: Thermo Scientific
16174 16175
CAPI16174EA 361.57 CAD
CAPI16174 CAPI16175
Pierce™ Firefly Luciferase Flash Assay Kit, Thermo Scientific
Assays Protein Assays
The Pierce™ Firefly Luciferase Flash Assay Kit provides reagents to measure Red firefly luciferase activity in whole cell producing a very strong flash signal which decays rapidly.
  • Highly sensitive detection of firefly luciferase activity
  • Amenable to automation
  • Contains a universal cell lysis buffer and optimized flash assay reagent
The bioluminescent signal produced by firefly luciferase results from the oxidation of D-Luciferin. The light output correlates with the amount of firefly protein expressed, which is proportional to the activity of the promoter for firefly expression.
Delivery information: Kit contains Firefly flash assay buffer (5 or 50 ml), D-Luciferin (3 or 30 mg) and 2X Cell lysis buffer (6 or 60 ml).
Caution: For research use only. Not for use in diagnostic procedures. It stored at 4 ℃.