Pierce™ Glutathione Spin Columns, Thermo Scientific
Supplier: Thermo Scientific
16106 16107 16108
CAPI16106EA 1053.13 CAD
CAPI16106 CAPI16107 CAPI16108
Pierce™ Glutathione Spin Columns, Thermo Scientific
Chromatography Columns
This Thermo Scientific Pierce GST Spin Purification Kit contains Pierce Glutathione Spin Columns and buffer components to provide a fast, easy-to-use format for purifying GST-fusion proteins from cellular lysates.
  • Glutathione is immobilized through its central sulfhydryl onto 6% crosslinked agarose resin
  • High capacity binds at least 40 mg of purified recombinant GST protein per milliliter of resin
  • Consistently purifies at least 10 mg of GST-tagged protein per milliliter of resin with greater than 90% purity
  • Resin can be reused at least five times without reduction in performance
Delivery information: Kit includes Pierce™ Glutathione spin columns, Equilibration/Wash Buffer, 125 mM Tris, Glutathione, and column plugs. The 0.2 mL kit also includes collection tubes.
Caution: For Research Use Only. Not for use in diagnostic procedures.