Pierce™ Glutathione Agarose Affinity Purification Media, Thermo Scientific
Supplier: Thermo Scientific
16100
16101
CAPI16100EA
698.46
CAD
CAPI16100
CAPI16101
Pierce™ Glutathione Agarose Affinity Purification Media, Thermo Scientific
Protein Purification
Protein Purification Resins
Thermo Scientific Pierce Glutathione Agarose is a high-capacity, high-performance resin for affinity purification of GST-tagged fusion proteins from cellular lysates
- Glutathione is immobilised through its central sulfhydryl onto 6% crosslinked agarose resin
- Binds at least 40 mg of purified recombinant GST protein per mL of resin
- Consistently purifies at least 10 mg of GST-tagged protein per mL of resin with greater than 90% purity
- Resin can be reused at least five times without reduction in performance
- Works well to purify GST-fusion proteins from bacterial lysates or with pre-purified GST-tagged proteins to pull down protein interactions
It is a crosslinked 6% beaded agarose, slurried in water with sodium azide.
Caution: For Research Use Only. Not for use in diagnostic procedures.