Pierce™ NeutrAvidin™ Coated Microplates, Thermo Scientific
Supplier: Thermo Scientific
15129 15123 15507 15509 15128 15400 15401 15402 15508 15510 15511 15513 15116 15117 15216 15217
PI15129EA 412.31 CAD
PI15129 CAPI-15123 CAPI-15507 CAPI-15509 CAPI15128 CAPI15400 CAPI15401 CAPI15402 CAPI15508 CAPI15510 CAPI15511 CAPI15512 CAPI15513 CAPI15116 CAPI15117 CAPI15216 CAPI15217
Pierce™ NeutrAvidin™ Coated Microplates, Thermo Scientific
Microplates Immunology Microplates and Strips
Thermo Scientific NeutrAvidin coated plates are designed for binding biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.
  • Unique plate-coating technology—results in high loading and binding capacity
  • NeutrAvidin Protein—coated plates have up to twice the binding capacity of typical plates
  • Broader dynamic range—extends the quantitative range so there is less need for dilutions
  • Save time—pre-blocked plates to reduce the number of assay steps
The proprietary plate-coating technology used to manufacture these Thermo Scientific Pierce Coated Plates yields high-capacity plates with a wider detection limit than regular binding capacity plates. The standard curve exhibits greater linearity for detecting small biotinylated molecules such as peptides and oligonucleotides, resulting in greater assay precision.
Caution: For Research Use Only. Not for use in diagnostic procedures.