Bruns Bone Curette, OR Grade, Sklar
Supplier: Sklar
40-7100 40-7112 40-7110 40-7114 40-7111 40-7113 40-7116 40-7000 40-7115 40-7118 40-7120 40-7200 40-7201 40-7204
10192-268EA 300.08 CAD
10192-268 10192-480 10192-486 10193-350 10193-786 10193-814 10194-236 10196-196 10196-198 10196-200 10196-202 10196-206 10196-208 10196-214
Bruns Bone Curette, OR Grade, Sklar
Curettes
Bruns Bone Curette is a debulking instrument commonly used in orthopedic procedures and neurosurgery. Also known as a Spratt Bone Curette. It is used to scrape away tissue and bone from the spine, bones, and joints.
  • Reusable and latex-free
  • Industry standard
  • Premium OR-grade
  • Non sterile
It is available in multiple sizes with various curvatures and tip styles to suit the surgical site and specifics of the procedure
This curette has a large, lightweight, hollow handle with longitudinal grooves and a thumb/finger rest for increased control.