2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH83630.400
BDH83630.400CS 1530.22 CAD
BDH83630.400
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
2,2,4-Trimethylpentane
Formula: (CH₃)₂CHCH₂C(CH₃)₃
Boiling Pt: 99.2 °C (1013 hPa)
Melting Pt: –107 °C
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) Min 99.5 %
Water (by KF coulometry) Max. 0.0100 %
Nonvolatile Residue Max. 0.0005 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Transmittance (220 nm) Min. 70 %
Transmittance (235 nm) Min. 80 %
Transmittance (255 nm) Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR