SGPF Series Sterile Powder Free Latex Gloves, CT International
Supplier: CT International
SGPF650 SGPF700 SGPF750 SGPF800 SGPF850 SGPF900 SGPF1000
56617-168CS 875.02 CAD
56617-168 56617-170 56617-172 56617-174 56617-176 56617-178 56617-180
SGPF Series Sterile Powder Free Latex Gloves, CT International
Gloves
SGPF Series are sterile powder free gloves. They are manufactured for outstanding strength, stretch and fit.
  • Textured surface and hand specific
  • Low protein, double chlorinated and contact sensitizers
  • Tapered, beaded cuff
  • Folded cuff for aseptic donning
  • Gamma irradiated sterilization minimum dose 2.5 mrads
They provides a textured surface and are double chlorinated reducing contact sensitizers and protein content. SGPF Series gloves are lot traceable.