PFXT Series Class 100 Powder Free Gloves, CT International
Supplier: CT International
PFXT201 PFXT202 PFXT203 PFXT204 PFXT101 PFXT102 PFXT103 PFXT104
56617-232CS 661.69 CAD
56617-232 56617-234 56617-236 56617-238 56617-240 56617-242 56617-244 56617-246
PFXT Series Class 100 Powder Free Gloves, CT International
Gloves
PFXT series are class 100 powder free gloves.
  • Manufactured for outstanding strength, stretch and fit
Offer a textured surface and are double chlorinated reducing contact sensitizers and protein content.
Certifications: PFXT Series gloves are ISO 5 Clean Room compatible or higher for controlled environments.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable
Latex/Latex-free Latex
Powdered/Powder-free Powder-free
Cleanroom class 100/ISO 5
ASTM No. D3578-05 