CHS Series Latex Water Washed Class 10 Gloves, CT International
Supplier: CT International
CHS600 CHS700 CHS650 CHS750 CHS800 CHS850 CHS1000 CHS900
56613-692CS 419.22 CAD
56613-692 56613-694 56613-739 56613-740 56613-741 56613-742 56613-746 56613-806
CHS Series Latex Water Washed Class 10 Gloves, CT International
Gloves
CHS Series are latex DI water washed Class 10 gloves.
  • Offers textured surface
They double chlorinated reducing contact sensitizers and protein content.
Like all our gloves, they are manufactured for outstanding strength, stretch and fit.
Certifications: CHS Series Class 10 Latex gloves ISO 4 Clean Room compatible or higher for controlled environments.