Boston Round Glass Bottles, Narrow Mouth, Clear, C&G Containers
Supplier: C&G Containers
LFB004900702 LFB032230000 LFB008900003 LFB004900100 LFB032900703 LFB032200000 LFB032900100 LFB008900000 LFB016200010 LFB008700403 LFB032900000 LFB0322100 LFB016900100 LFB004200010 LFB032900903 LFB016900000 LFB004900000 LFB008200012 LFB016100000 LFB0169002 LFB0049005 LFB008900500 LFB004400010 LAB032200010 LFB0029005 LFB008200000 LFB008100402 LFB008700001 LFB0329005
71000-750CS 256.49 CAD
71000-750 71000-754 71000-732 71000-730 30617-858 10146-768 30617-856 10146-732 10146-752 71000-604 71000-722 71000-728 71000-748 71000-726 71000-760 30617-862 71000-740 89239-474 71000-140 10146-764 10146-766 10146-762 30621-156 71000-736 71000-758 71000-756 89106-732 71000-718 71000-738
Boston Round Glass Bottles, Narrow Mouth, Clear, C&G Containers
Bottles Glass Bottles
These glass jars are used primarily for the collection of liquid media, for testing semi-volatiles, pesticides, PCBs, diesel range organics, TPH, metals, and phenolics.
Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), D1 (Depyrogenation-like process), and L3 (no wash).
Secondary uses include containment and archiving. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity 'contain' volumes may vary.