Rein Round Curette, OR Grade, Sklar
Supplier: Sklar
06-4059 06-4060 06-4061
10194-878EA 287.94 CAD
10194-878 10194-880 10194-882
Rein Round Curette, OR Grade, Sklar
Curettes
Sklar® Rein Round Curette is a reusable, stainless steel, OR grade instrument.
  • Used in dermatology
  • General surgery
  • Physician
  • Podiatry specialties
This product measures 5.5” in length, straight, with round tip.