Round-Bottom Three-Neck Flasks, Angled Side Necks, Heavy Wall, Ace Glass Incorporated
Supplier: Ace Glass
6948-240 6948-262 9465-207 6948-208 9465-211 6948-224 6948-239 9466-224 6948-258 6948-236 6948-242 6948-216 6948-205 6948-267 6948-203 6948-237 6948-212 6948-263 6948-226 6948-207 9465-209 6948-234 6948-232 9466-220 9466-214 6948-260 9466-210 9466-208 6948-241 6948-238 6948-222 6948-235 6948-220 9466-212 6948-261 9466-206 9465-213 9465-204 6948-227
89085-112EA 278.8 CAD
89085-112 89057-124 89056-838 89074-768 89074-728 89056-860 89056-840 89056-862 89056-880 89056-868 89056-824 89056-846 89085-110 89056-864 89056-820 89056-822 89056-844 89074-696 89074-730 89057-130 89074-694 89056-828 89074-698 89074-732 89057-132 89057-154 89057-216 89074-716 89085-108 89056-476 89056-850 89056-856 89056-878 89056-834 89074-692 89057-192 89056-874 89056-876 89056-854
Round-Bottom Three-Neck Flasks, Angled Side Necks, Heavy Wall, Ace Glass Incorporated
Flasks Round Bottom Flasks
These round-bottom flasks have angled side necks and [ST] outer joints
The outer joints are tooled with reinforcement rings (full-sized joints only) for added strength and stability. The flare-out directly below the center joint facilitates cleaning of the flask and enables stirring blades to be easily removed. Fabricated from borosilicate glass. Heavy wall flasks feature walls that are approximately 30% heavier than standard wall flasks.