Disposable Ear Curette, Floor Grade, Sklar
Supplier: Sklar
96-1000 96-1010 96-1015 96-1020 96-1030 96-1021 96-1005 96-1031 96-1006 96-1016 96-1001 96-1011 96-1035
10185-248PK 48.53 CAD
10185-248 10185-250 10185-252 10185-254 10185-256 10192-304 10192-402 10193-298 10193-648 10193-694 10193-708 10193-822 10193-876
Disposable Ear Curette, Floor Grade, Sklar
Curettes
The ear curette is manufactured from a cold sterilizable polymer.
  • Firm, and pliable
  • Non-autoclavable
  • Non sterile
These curettes eliminate the risk of cross infection.
Ordering information: The ear curette assortment includes 10 curettes of each type.