Barnhill Adenoid Curette, OR Grade, Sklar
Supplier: Sklar
70-9200 70-9201 70-9202 70-9203 70-9204
10198-570EA 692.06 CAD
10198-570 10198-572 10198-574 10198-576 10198-578
Barnhill Adenoid Curette, OR Grade, Sklar
Curettes
Barnhill adenoid curette is a debulking instrument used primarily in adenoidectomy procedures.
  • Curved and rectangular
  • Fenestrated tip
The Barnhill Adenoid Curette is similar to the Beckman adenoid curette; however, the working end of the Beckman curette is slightly wider and shorter, resembling more of a square shape. It is also slightly longer in overall length.
It has a sharp inner blade that is used to scrape away adenoids or tonsils. It is available with a variety of blade widths.