Sims Uterine Curette, OR Grade, Sklar
Supplier: Sklar
90-5424 90-5425 90-5426 90-5500 90-5421 90-5422 90-5423
10197-986EA 301.57 CAD
10197-986 10197-988 10197-990 10198-006 10198-008 10199-292 10199-294 10199-296
Sims Uterine Curette, OR Grade, Sklar
Curettes
Sims Uterine Curette is an instrument used for removing tissue from the lining of the uterus.
  • Hollow with vertical grooves handle
  • Reusable and non sterile
  • Latex-Free
It may be used to scrape and remove growths or tissue to be biopsied. It may also be used as part of a D&C procedure. The teardrop shaped, fenestrated, loop tip is sharp and available in various sizes. The shaft is malleable so that it can be manipulated to an angle suitable for both user and patient needs.