Human Single Tissue Single Species Blot, Normal and Tumor Tissue, G-Biosciences
Supplier: G-Biosciences
TB92 TB93 TB94 TB95 TB96 TB97 TB98 TB100
CA95029-402EA 2878.55 CAD
CA95029-402 CA95029-404 CA95029-406 CA95029-408 CA95029-410 CA95029-412 CA95029-414 CA95029-418
Human Single Tissue Single Species Blot, Normal and Tumor Tissue, G-Biosciences
Blotting Kits
G-Biosciences offers a selection of ready-to-screen, human, single tissue blots, from both normal and tumor tissues.
  • Blots are ready to be blocked and probed with the antibodies of choice
  • Each blot contains two protein lanes, a protein marker lane and a lane with GenLysate™ sample
G-Biosciences premade western blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
Ordering information: Accessories available.