Human Single Tissue Single Species Blot, Tumor Tissue, G-Biosciences
Supplier: G-Biosciences
TB54 TB72 TB73 TB74 TB75 TB76 TB77 TB78 TB80 TB81 TB82 TB83 TB84 TB85 TB86 TB87 TB88 TB89 TB90 TB91
CA82023-238EA 5912.45 CAD
CA82023-238 CA95029-362 CA95029-364 CA95029-366 CA95029-368 CA95029-370 CA95029-372 CA95029-374 CA95029-378 CA95029-380 CA95029-382 CA95029-384 CA95029-386 CA95029-388 CA95029-390 CA95029-392 CA95029-394 CA95029-396 CA95029-398 CA95029-400
Human Single Tissue Single Species Blot, Tumor Tissue, G-Biosciences
Blotting Kits
G-Biosciences offers a selection of ready-to-screen, human, single tissue blots, from tumor tissues.
  • Blots are ready to be blocked and probed with the antibodies of choice
  • Each blot contains two protein lanes, a protein marker lane and a lane with GenLysate™ sample
G-Biosciences premade western blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using ourMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable