Wide Mouth Glass Jars, Clear, C&G Containers
Supplier: C&G Containers
LFQ032200010 LFQ032210000 LFQ032900000
71000-802CS 170.62 CAD
71000-802 71000-804 71000-806 71000-808 71000-810
Wide Mouth Glass Jars, Clear, C&G Containers
Jars General Use Jars
These glass jars are used primarily for the collection of liquid media, soil, sediments, and slurry media for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics
Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.