MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80075-HF
80075-PD
80075-SB
80075-SE
80075-Y
80075-AS
80075-BE
80075-C
80075-PT
80075-TE
80075-CU
80075-NB
80075-RH
80075-SC
80075-AG
80075-AL
80075-MN
80075-SI
80075-CO
80075-GE
80075-SN
80075-CD
80075-RU
80075-TL
80075-ZR
80075-FE
80075-MG
80075-TA
80075-TI
80075-BAS
80075-BAS-FC
100495-174EA
824.55
CAD
100495-174
100495-200
100495-210
100495-214
100495-232
100495-140
100495-154
100495-158
100495-202
100495-222
100495-166
100495-184
100495-206
100495-212
100495-136
100495-138
100495-180
100495-216
100495-162
100495-172
100495-218
100495-160
100495-208
100495-226
100495-236
100495-168
100495-178
100495-220
100495-224
102099-030
102099-032
MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Standards
Colour Reference and Spectroscopy Standards
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with a location map for standard identification. Grains of standard materials are available and they measure approximately 2 x 2 mm in size. They are supplied with fully authenticated certificates of analysis.