Atlantic City (AC) Round Glass Bottles, Clear, C&G Containers
Supplier: C&G Containers
LFA004400010 LFA004900000 LFA008200010 LFA008900000 LFA016200010 LFA016900000 LFA008200000
71000-700CS 1002.27 CAD
71000-700 71000-702 71000-704 71000-706 71000-708 71000-710 71000-712 71000-714 71000-716 89125-402
Atlantic City (AC) Round Glass Bottles, Clear, C&G Containers
Bottles Glass Bottles
These jars are used primarily for the collection of of soil, sediments, and slurry media for testing Semi-volatiles, pesticides, PCBs, diesel range Organics, TPH, metals, and phenolics.
Pre-cleaned and certified products contain the certificate of analysis and cleaning protocol form in the product case.
Pre-cleaned and certified products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), and L3 (no wash).
Secondary uses include containment, bench vessels, and archiving. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity 'contain' volumes may vary.