First Aid and Spill Response Signs without Header, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
M249P M249R M346PB M346RB M441P M441R M441RB M442P M442R M442RB M458RB M459RB M501AB M501P M501R M501RB M65P M65PP M65R M81PB M81RB SPSA172P SPSA172R SPSA173P SPSA173R
94017-030EA 33.21 CAD
94017-030 94017-032 94017-038 94017-040 94017-090 94017-092 94017-242 94017-244 94017-246 94017-248 94017-250 94017-252 94017-304 94017-306 94017-316 94017-318 94017-320 94017-322 94017-372 94017-374 94017-376 94017-472 94017-474 94021-868 94021-870 94021-900 94021-902 94021-904 94021-906
First Aid and Spill Response Signs without Header, National Marker
Signs
Alert your employees to the location of first aid and emergency response stations.
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes