Mayo Glass Bottles, Clear, C&G Containers
Supplier: C&G Containers
LFM012900000 LFM008400010 LFM012400010 LFM032200010 LFM008900000 LFM016900000 LFM008308100 LFM016200000 LFM032900000
71000-782CS 117.2 CAD
71000-782 71000-780 71000-792 71000-786 71000-784 89137-860 71000-788 71000-778 71000-790
Mayo Glass Bottles, Clear, C&G Containers
Bottles Glass Bottles
These glass jars are used primarily for the collection of soil and slurry media for testing Semi-volatiles, Pesticides, PCBs, diesel range organics, TPH, metals, and phenolics.
Pre-cleaned and certified products contain the certificate of analysis and cleaning protocol form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for pre-cleaned and certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (nitric acid wash), B1 (deionized water wash), C1 (hydrochloric acid wash), and L3 (no wash). Some items are Nitrogen-purged.
Secondary uses include containment, bench vessels, and archiving. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity 'contain' volumes may vary.