Wide Mouth Glass Jugs, Clear, C&G Containers
Supplier: C&G Containers
LFW320900000 LFW128900000 LFW320200010 LFW064200000 LFW064900000 LFW128200010 LFW064200010 LFW128100000
89125-410CS 132.5 CAD
30621-208 71000-871 89125-410 71000-873 71000-862 71000-854 71000-866 71000-858
Wide Mouth Glass Jugs, Clear, C&G Containers
Bottles Carboys
These glass jugs are used primarily for the collection of benthic organisms and the collection of water samples for mulitple analyte testing.
Pre-cleaned and certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols for these jugs include; A1 (nitric acid wash), B1 (deionized water wash), C1 (hydrochloric acid wash), and L3 (no wash).
Secondary uses include containment and archiving. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity 'contain' volumes may vary.