Mammalian GenLysate™ Human Total Protein Lysates, Tumor Tissue, G-Biosciences
Supplier: G-Biosciences
TLH-01 TLH-03 TLH-06 TLH-07 TLH-09 TLH-11 TLH-12 TLH-13 TLH-14 TLH-15 TLH-16 TLH-17 TLH-18 TLH-19 TLH-20
CA82022-288EA 730.88 CAD
CA82022-288 CA82022-292 CA82022-296 CA82022-300 CA82022-304 CA82022-308 CA82022-312 CA82022-316 CA82022-320 CA82022-324 CA82022-328 CA82022-332 CA82022-336 CA82022-340 CA82022-344
Mammalian GenLysate™ Human Total Protein Lysates, Tumor Tissue, G-Biosciences
Cells
G-Biosciences' GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes.
  • Extracted from the tissue or cell of interest
  • Ready for use
  • Available in a wide variety of different species and tissues
G-Biosciences' GenLysates™ are total protein tissue and cell lysates that are extracted from the tissue or cell of interest
The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris HCl and 2% SDS with G-Biosciences Protease ARREST™ protease inhibitor cocktail. This minimizes proteolytic damage to the proteins.
G-Biosciences GenLysate™ total protein lysates are offered as a 150 µg lyophilized protein per vial and, following reconstitution in DI water, are ready for use. G-Biosciences offers GenLysate™ in a wide variety of different species and tissues. Species include: human (both normal tissue and tumor tissue lysates available), mouse, rat, macaque (primate), rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.
To inhibit proteolysis, the GenLysates™ are prepared in a lysis buffer containing a complete protease inhibitor cocktail.