Metzenbaum-Nelson Bariatric Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
55-9612 55-9614
10197-686EA 709.23 CAD
10197-686 10197-688
Metzenbaum-Nelson Bariatric Dissecting Scissors, OR Grade, Sklar
Scissors
Metzenbaum-Nelson Dissecting Scissors are designed for dissecting and cutting tissue.
They tend to be longer than standard Metzenbaum scissors, especially for use in thoracic procedures.
Not recommended for cutting sutures, drains, or heavy tissue.