Polyester Printer Labels, Brady
Supplier: Brady
BMP®50
143386 143311 143347 MC-1250-461 143325 143326 143301 143343 MC-500-492 143303 143342 M-112-490 143307 131575 143346 143282 134100 143291 143378 143345 143322 143374 143316 143317 143341 143292 143357 143269 M-82-492 134095 M-131-492 143366 143283 143319 M-156-492 143284 M-117-492 143344 143382 143288 143318 143286 143323 M-72-461 143233 143310 134099 143302 134101 143398 143394 M-97-481 143290 143300 143402 143293 143340 143339 143327 M-133-427 143313 143356 131591 143312 143299 143295 143362 M-118-492 143294 143355 143362 143390 143315 M-120-492 143297 143256 143314 143324 MC-375-461 143287 143370 143308 143298 143289 143339 143309
CA89208-544EA 92.74 CAD
89208-538 89208-456 CA89208-544 89208-458 89219-624 89219-590 89219-592 89219-594 89219-596 89208-540 89208-462 89208-586 89208-464 89208-580 89208-460 89208-526 89208-448 89208-528 89208-522 89208-444 89208-488 89208-568 89208-524 89208-446 CA10748-142 490006-148 89208-490 89208-452 89208-496 89208-498 89208-454 89208-530 89208-574 89208-492 89208-494 89208-514 10796-512 89208-510 10796-158 89208-512 89208-556 CA89208-520 490006-150 490006-194 490006-196 490006-152 89219-604 490006-198 490006-154 490006-156 490006-158 CA89234-496 89208-562 89208-484 89208-520 89208-442 89208-486 10796-486 89208-504 89208-426 10796-484 89208-506 10796-242 89208-544 89208-500 89208-466 89208-502 89208-468 89219-598 490006-160 10796-160 490006-162 89219-634 10796-482 10796-240 89208-508 10796-162 490006-202 490006-200 89208-474 89208-550 89234-496 89208-476 89208-470 89208-472 89208-592
Polyester Printer Labels, Brady
Labels
B-461: Self-laminating polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-490: Durable polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
B-492: Durable Freezerbondz® polyester markers are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
B-481: StainerBondz™ super chemical resistant polyester is designed for use in the slide staining process and other laboratory ID which requires superior chemical resistance.
B-482: Tissue cassette polyester is designed for tissue cassette labeling and identification in conjunction with the BSP™31 Label Attachment System from Brady®.
B-488: High performance polyester withstands numerous solvents and variable temperatures when applied to various surfaces including stainless steel and polypropylene.
B-498: Permanent or repositionable, once applied, CleanLift® series vinyl cloth is there to stay unless you want to change it.
B-427: Self-laminating vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
B-432: Rough surface polyester labels are ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
B-595: This durable, low-shrink vinyl with ultra-aggressive adhesive is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
B-351: Tamper-resistant vinyl is designed to fracture easily in order to show signs of product tampering and to prevent on-piece label removal.