ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 68 to 136, Thermco Products
Supplier: Thermco
ACC068CFC ACC068FFC ACC069CFC ACC069FFC ACC070CFC ACC070FFC ACC073CBLSFC ACC077FFC ACC078FFC ACC079FFC ACC080FFC ACC081FFC ACC097CFC ACC097FFC ACC108FFC ACC109FFC ACC110FFC ACC111CFC ACC112CFC ACC113CFC ACC113FFC ACC114CFC ACC116CFC ACC117CFC ACC118CFC ACC118FFC ACC120CFC ACC121CFC ACC123CFC ACC124CFC ACC125CFC ACC126CFC ACC126FFC ACC127CFC ACC128CFC ACC128FFC ACC129CFC ACC129FFC ACC130CFC ACC130FFC ACC132CFC ACC136CFC ACC136FFC ACC098CFC ACC098FFC
10747-592EA 1391.71 CAD
10747-592 10747-594 10747-596 10747-598 10747-600 10747-602 10747-608 10747-610 10747-612 10747-614 10747-624 10747-626 10747-628 10747-630 10747-632 10747-678 10747-680 10747-682 10747-684 10747-686 10747-688 10747-690 10747-692 10747-694 10747-696 10747-698 10747-702 10747-704 10747-706 10747-708 10747-714 10747-716 10747-720 10747-722 10747-724 10747-726 10747-728 10747-730 10747-732 10747-734 10747-736 10747-738 10747-740 10747-742 10747-744 10747-754 10747-756 10747-850 10747-852
ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 68 to 136, Thermco Products
Thermometers Analog Thermometers
These high precision liquid-in-glass mercury thermometers are hand blown, which bear graduations, numbers and letters etched into the glass and filled with and inert gas above the mercury column
ASTM thermometers are used for a variety of specified tests which quantify and qualify repeatable test results in the fields of oil & petroleum testing.
These thermometers were designed for specific testing and can be calibrated to your specific requirements.
Certifications: A NIST Traceable Certificate of Calibration from Thermco Products ISO/IEC accredited laboratory at the specified certification points.
Ordering information: Accessories not included.
Caution: WARNING—Mercury has been designated by EPA and many state agencies as a hazardous material that can cause central nervous system, kidney and liver damage. Mercury, or its vapor, may be hazardous to health and corrosive to materials. Caution should be taken when handling mercury and mercury containing products. See the applicable product Safety Data Sheet (SDS) for details.