Lab Worktables, Stationary, Metro™
Supplier: Metropolitan Wire
LTS30IS LTS48IS LTS30UIS LTS36UIS LTS48UIS LTS60UIS LTS30S LTS36S LTS48S LTS60S LTS30US LTS36US LTS48US LTS60US LTS30PB LTS36PB LTS48PB LTS60PB LTS30PG LTS36PG LTS48PG LTS60PG LTS30UPB LTS36UPB LTS48UPB LTS60UPB LTS36IS LTS60IS LTS30UPG LTS36UPG LTS48UPG LTS60UPG
CA97002-700EA 3335.07 CAD
CA97002-700 CA97002-704 CA97002-716 CA97002-718 CA97002-720 CA97002-722 CA97002-732 CA97002-734 CA97002-736 CA97002-738 CA97002-748 CA97002-750 CA97002-752 CA97002-754 CA97002-764 CA97002-766 CA97002-768 CA97002-770 CA97002-796 CA97002-798 CA97002-800 CA97002-802 CA97002-812 CA97002-814 CA97002-816 CA97002-818 CA97005-904 CA97005-906 CA97005-908 CA97005-910 CA97005-912 CA97005-914
Lab Worktables, Stationary, Metro™
Furniture Tables and Workbenches
Stainless steel lab worktables provides corrosion-proof performance. Stainless steel provides excellent protection in corrosive environments.
  • Tables feature smooth, easy-to-clean surfaces
  • Tops are made of Type 304 stainless steel or Trespa® phenolic resin
  • Phenolic resin tops are compression molded of organic fiber-reinforced phenolic core with an integrally cured thermoset resin surface
  • Phenolic resin tops are ideal for moderate to high chemical use and have excellent abrasion resistance and impact resistance
  • Tops with backsplash feature a 10.2 cm (4")
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable