Round-Bottom Three-Neck Flasks, Vertical Necks, Heavy Walls, Ace Glass Incorporated
Supplier: Ace Glass
6944-203 6944-204 6944-205 6944-206 6944-210 6944-216 6944-218 6944-220 6944-222 6944-223 6944-224 6944-225 6944-226 6944-228 6944-229 6944-230 6944-231 6944-238 6944-240 6944-242 6944-244 6944-245 6944-246 6944-247 6944-248 6944-249 6944-250 6944-254 6944-256 6944-257 6944-258 6944-259 6944-260 6944-262 6944-264 6944-266 6944-267 6944-268 6944-294 6944-297 6944-207
89056-608EA 360.94 CAD
89056-608 89056-610 89056-612 89056-614 89056-616 89056-618 89056-620 89056-624 89056-626 89056-628 89056-630 89056-632 89056-634 89056-636 89056-638 89056-640 89056-642 89056-644 89056-648 89056-650 89056-652 89056-654 89056-656 89056-658 89056-660 89056-662 89056-666 89056-668 89056-670 89056-672 89056-674 89056-676 89056-680 89056-682 89056-684 89056-686 89056-688 89056-690 89056-694 89056-696 89083-882
Round-Bottom Three-Neck Flasks, Vertical Necks, Heavy Walls, Ace Glass Incorporated
Flasks Round Bottom Flasks
Round-bottom flasks with three vertical necks and [ST] outer joints
The outer joints are tooled with reinforcement rings (full-sized joints only) for added strength and stability. The flare-out directly below the center joint facilitates cleaning of the flask and enables stirring blades to be easily removed. Fabricated from borosilicate glass. Heavy wall flasks feature walls that are approximately 30% heavier than standard wall flasks.